Acetonitrile ≥99.9%, B&J Brand™ ACS, meets UV Specifications for HPLC, for spectrophotometry carbonyl-free, Burdick & Jackson™

Supplier: HONEYWELL INTERNATIONAL INC MX
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

018-4 018-1L
BJ018-4CS 2081 USD
BJ018-4 BJ018-1L
Acetonitrile ≥99.9%, B&J Brand™ ACS, meets UV Specifications for HPLC, for spectrophotometry carbonyl-free, Burdick & Jackson™
Acetonitrile

Acetonitrile carbonyl-Free, Meets UV Specifications.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonyl Compounds (as HCHO) 10 ppb max.
Water by Karl Fischer Titration 0.01 % max.
GC Analysis 99.9 % min.
HPLC Gradient @ 215 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 1 mg/L max.
UV Absorbance @ 200 nm 0.05 AU max.
UV Absorbance @ 210 nm 0.04 AU max.
UV Cutoff 190 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR