Test Strips, Peracetic acid, Dosatest, VWR Chemicals BDH®
Supplier: VWR International
91340.AVA 91341.AVA 91342.AVA
BDH85436.601EA 106.47 USD
BDH85436.601 BDH85437.601 BDH85438.601
Test Strips, Peracetic acid, Dosatest, VWR Chemicals BDH®
Environmental Test Kits
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection process. Semi-quantitative results can be obtained between 10 and 120 seconds.
Color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Envase: Container with 100 test strips 6×95 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable