Total Ionic Strength Adjustment Buffer Solution (TISAB), VWR Chemicals BDH®
Supplier: VWR International
BDH7492-4
BDH7492-4CS 1473.84 USD
BDH7492-4
Total Ionic Strength Adjustment Buffer Solution (TISAB), VWR Chemicals BDH®
Buffers Ionic Strength Adjustment Buffers
ASTM formulation D 1179 with deionized water, acetic acid, sodium chloride and sodium citrate.