Sulphuric acid 10 N aqueous solution zA, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7259-1
BDH7259-1CS 673.87 USD
BDH7259-1
Sulphuric acid 10 N aqueous solution zA, VWR Chemicals BDH®
Sulfuric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 9.95 - 10.05 N
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR