Sulfuric acid 0.1 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7230-1 BDH7230-4 BDH7230-7
BDH7230-1CS 523.34 USD
BDH7230-1 BDH7230-4 BDH7230-7
Sulfuric acid 0.1 N in aqueous solution, VWR Chemicals BDH®
Sulfuric acid

Meets ASTM E200-91 and AOAC 890-01 standards. Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 0.0995 - 0.1005 N
Shelf-life 2 years

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR