pH Reference Standard Buffers, VWR Chemicals BDH®

Supplier: BDH

BDH01880-55E BDH50000-20F BDH50120-20F BDH50160-20PK BDH50280-20F BDH50360-20F BDH50420-20F BDH50440-20PK BDH50680-20F BDH50700-20PK BDH50900-20F BDH50960-20F BDH01900-20F BDH01920-55E BDH01940-20F BDH01960-55E BDH01980-20F BDH50020-500A BDH50040-20F BDH50060-500A BDH50080-4A BDH50100-500A BDH50140-500A BDH50180-500A BDH50200-4A BDH50220-4F BDH50240-500A BDH50260-4A BDH50300-4A BDH50320-500A BDH50340-500A BDH50380-500A BDH50400-500A BDH50460-500A BDH50480-4A BDH50500-4F BDH50520-500A BDH50540-4A BDH50560-20F BDH50580-500A BDH50600-500A BDH50620-500A BDH50640-4A BDH50660-500A BDH50720-500A BDH50740-4A BDH50760-4F BDH50780-500A BDH50800-4A BDH50820-20F BDH50840-500A BDH50860-500A BDH50880-4A BDH50920-500A BDH50940-4A BDH50980-4A BDH51000-6PK
BDH0188-55GLEA 1268.96 USD
BDH0188-55GL BDH5000-20L BDH5012-20L BDH5016-20ML BDH5028-20L BDH5036-20L BDH5042-20L BDH5044-20ML BDH5068-20L BDH5070-20ML BDH5090-20L BDH5096-20L BDH0190-20L BDH0192-55GL BDH0194-20L BDH0196-55GL BDH0198-20L BDH5002-500ML BDH5004-20L BDH5006-500ML BDH5008-4L BDH5010-500ML BDH5014-500ML BDH5018-500ML BDH5020-4L BDH5022-4L BDH5024-500ML BDH5026-4L BDH5030-4L BDH5032-500ML BDH5034-500ML BDH5038-500ML BDH5040-500ML BDH5046-500ML BDH5048-4L BDH5050-4L BDH5052-500ML BDH5054-4L BDH5056-20L BDH5058-500ML BDH5060-500ML BDH5062-500ML BDH5064-4L BDH5066-500ML BDH5072-500ML BDH5074-4L BDH5076-4L BDH5078-500ML BDH5080-4L BDH5082-20L BDH5084-500ML BDH5086-500ML BDH5088-4L BDH5092-500ML BDH5094-4L BDH5098-4L BDH5100-PK
pH Reference Standard Buffers, VWR Chemicals BDH®
Buffers pH Calibration Buffers

BDH pH buffers are available as either colorless or colored solutions.


  • Traceable to NIST Standards reference materials
  • Calibrated in an ISO 17025 certified laboratory using certified Standards
  • Store at room temperature


Información para pedidos: Buffer set BDH5100-PK comes with two 500 mL bottles each of the pH 4.00 red buffer, pH 7.00 yellow buffer, and pH 10.00 blue buffer.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR