pH Reference Standard Buffers, VWR Chemicals BDH®

Supplier: BDH

BDH01880-55E BDH50000-20F BDH50120-20F BDH50160-20PK BDH50280-20F BDH50360-20F BDH50420-20F BDH50440-20PK BDH50680-20F BDH50700-20PK BDH50900-20F BDH50960-20F BDH01900-20F BDH01920-55E BDH01940-20F BDH01960-55E BDH01980-20F BDH50020-500A BDH50040-20F BDH50060-500A BDH50080-4A BDH50100-500A BDH50140-500A BDH50180-500A BDH50200-4A BDH50220-4F BDH50240-500A BDH50260-4A BDH50300-4A BDH50320-500A BDH50340-500A BDH50380-500A BDH50400-500A BDH50460-500A BDH50480-4A BDH50500-4F BDH50520-500A BDH50540-4A BDH50560-20F BDH50580-500A BDH50600-500A BDH50620-500A BDH50640-4A BDH50660-500A BDH50720-500A BDH50740-4A BDH50760-4F BDH50780-500A BDH50800-4A BDH50820-20F BDH50840-500A BDH50860-500A BDH50880-4A BDH50920-500A BDH50940-4A BDH50980-4A BDH51000-6PK
BDH0188-55GLEA 1268.96 USD
BDH0188-55GL BDH5000-20L BDH5012-20L BDH5016-20ML BDH5028-20L BDH5036-20L BDH5042-20L BDH5044-20ML BDH5068-20L BDH5070-20ML BDH5090-20L BDH5096-20L BDH0190-20L BDH0192-55GL BDH0194-20L BDH0196-55GL BDH0198-20L BDH5002-500ML BDH5004-20L BDH5006-500ML BDH5008-4L BDH5010-500ML BDH5014-500ML BDH5018-500ML BDH5020-4L BDH5022-4L BDH5024-500ML BDH5026-4L BDH5030-4L BDH5032-500ML BDH5034-500ML BDH5038-500ML BDH5040-500ML BDH5046-500ML BDH5048-4L BDH5050-4L BDH5052-500ML BDH5054-4L BDH5056-20L BDH5058-500ML BDH5060-500ML BDH5062-500ML BDH5064-4L BDH5066-500ML BDH5072-500ML BDH5074-4L BDH5076-4L BDH5078-500ML BDH5080-4L BDH5082-20L BDH5084-500ML BDH5086-500ML BDH5088-4L BDH5092-500ML BDH5094-4L BDH5098-4L BDH5100-PK
pH Reference Standard Buffers, VWR Chemicals BDH®
Buffers pH Calibration Buffers

BDH pH buffers are available as either colorless or colored solutions.


  • Traceable to NIST Standards reference materials
  • Calibrated in an ISO 17025 certified laboratory using certified Standards
  • Store at room temperature


Información para pedidos: Buffer set BDH5100-PK comes with two 500 mL bottles each of the pH 4.00 red buffer, pH 7.00 yellow buffer, and pH 10.00 blue buffer.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR