Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

005700-80601 005700-80602 005700-80603 005700-80401 005700-03901 005700-22001 005700-80604
BDH3092-500MLPCS 557.99 USD
BDH3092-500MLP BDH3094-2.5LG BDH3096-2.5LPC BDH3098-3.8LP BDH3100-19L BDH3102-201L BDH3093-2.2LP
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Acetic acid

Clear, colorless liquid.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3COOH) 99.7% min.(ACS)/99.5-100.5%(USP/FCC)
Acetic Anhydride 0.01% max.(ACS)
Chloride (as Cl) To Pass Test(USP)
Chloride (Cl) 1 ppm max.(ACS)
Color (APHA) 10 max.(ACS)
Congealing Temperature 15.6 C min.(USP/FCC)
Dilution Test To pass test(ACS)
Heavy Metals (as Pb) 0.5 ppm max.(ACS)
Identification: acetate To Pass Test(USP/FCC)
Iron (Fe) 0.2 ppm max.(ACS)
Lead (Pb) 0.5 mg/kg max.(FCC)
Nonvolatile residue 1.0mg max.(USP)/0.005% max.(FCC)
Oxidizable substances To pass test(USP/FCC)
Residue After Evaporation 0.001% max.(ACS)
Substances Reducing Dichromate To pass test (ACS)
Substances Reducing Permanganate To pass test (ACS)
Sulfate 1 ppm max.(ACS)
Sulfate (SO4) To Pass Test(USP)
Titrable Base 0.0004 meq/g max.(ACS)
Manufacture Date Record Actual
Expiration date Record Actual


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR