Hydrochloric acid solution_SC 0.600 N zA, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH3021-20L
BDH3021-20LEA 318.9 USD
BDH3021-20L
Hydrochloric acid solution_SC 0.600 N zA, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Assay (HCl) 0.5990 - 0.6010N
Color (APHA) 10 max.
Manufacture Date Record


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR