n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: high purity solvents

Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH24539.400
BDH24539.400CS 1516.6 USD
BDH24539.400
n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4
Melting Pt: -90.6
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation Residue Max. 10 ppm
Water Max. 100 ppm
Transmittance (210 nm) Min. 50 %
Transmittance (220 nm) Min. 80 %
Transmittance (245 nm) Min. 98 %
Conforms to BDH 15288 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR