Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

BDH20067.400
BDH20067.400CS 657.18 USD
BDH20067.400
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) (Corrected for Water) Min. 99.5 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Residue after evaporation Max. 5 ppm
Water (by KF coulometry) Max. 0.5 %
Transmittance (345 nm) Min. 50 %
Transmittance (350 nm) Min. 80 %
Transmittance (360 nm) Min. 98 %

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental Certification Chem21 Solvent selection guide recommended after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR