1-Methylimidazole (NMI) 20% in anhydrous THF for biosynthesis Cap B, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

CB3979-014 CB3979-001 CB3979-051
BDH0608-2.5LCS 4837.96 USD
BDH0608-2.5L BDH0608-4L BDH0608-56L
1-Methylimidazole (NMI) 20% in anhydrous THF for biosynthesis Cap B, VWR Chemicals BDH®
1-Methylimidazole (NMI)

BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.


  • Custom Blends and Bulk Sizes available upon request.

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198
Melting Pt: -2
Flash Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
UN: 2924
ADR: 3 (8) II

Order Now

Specification Test Results

Description (Clear liquid free of suspended matter) Pass
1-Methyl imidazole 15.0 - 17.0%
Tetrahydrofuran 83.0 - 85.0%
Color (at time of packaging) <10 APHA
Water Max 150 ppm
1-Methyl imidazole (component purity) Min 99.0%
Tetrahydrofuran (component purity) Min 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR