3-Amino-5-bromopyridine-2-carbonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-Amino-5-bromopicolinonitrile

A185306-1G
ALA185306-1GEA 575.61 USD
ALA185306-1G
3-Amino-5-bromopyridine-2-carbonitrile ≥97%
3-Amino-5-bromopyridine-2-carbonitrile
Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375025
CAS Number: 573675-27-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14282828c34c4a67f3a5ffec4760b4faada7ba36&vid=14282828c34c4a67f3a5ffec4760b4faada7ba36&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR