4-Aminobenzamidoxime 97

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Aminobenzamide oxime, 4-Amino-N'-hydroxybenzimidamide

A169216-1G
ALA169216-1GEA 48.36 USD
ALA169216-1G
4-Aminobenzamidoxime 97
4-Aminobenzamidoxime
Formula: C₇H₉N₃O
MW: 151.17
Melting Pt: 167-171°C
Storage Temperature: Ambient
MDL Number: MFCD08061151
CAS Number: 277319-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a17da3903522e0aab8c93b9dc817ddc00b157dd8&vid=a17da3903522e0aab8c93b9dc817ddc00b157dd8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR