Acridine Mutagen ICR 191 ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A167847-100MG
ALA167847100MGEA 1565.51 USD
ALA167847100MG
Acridine Mutagen ICR 191 ≥95%
Acridine Mutagen ICR 191
Formula: C₁₉H₂₁Cl₂N₃O ·2HCl
MW: 451.22 g/mol
Storage Temperature: Refrigerator
CAS Number: 17070-45-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09bad3a79ffc91c95ebb7445eec388aaabddbe3c&vid=09bad3a79ffc91c95ebb7445eec388aaabddbe3c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR