Azido PEG 95
Supplier: ALADDIN SCIENTIFIC
Synonyms: mPEG-N3
A163311-1G A163311-5G
ALA163311-1GEA 539.51 USD
ALA163311-1G ALA163311-5G
Azido PEG 95
Azido PEG
Azide/azido functionalized methoxyl polyethylene glycol (mPEG-N3) is a monofunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous solution catalyzed by copper. It can also be reduced into amine group easily. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.