Azido PEG
Supplier: ALADDIN SCIENTIFIC
Synonyms: mPEG-N3
A163277-100MG A163277-500MG
ALA163277100MGEA 262.11 USD
ALA163277100MG ALA163277500MG
Azido PEG
Azido PEG
Azide/azido functionalized polyethylene glycol (N3-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous solution catalyzed by copper. It can also be reduced into amine group easily while another functional group can be react with other molecules or functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.