2-Acetamido-6-nitrobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetamido-6-nitrobenzoic Acid, 2'-Carboxy-3'-nitroacetanilide, N-Acetyl-6-nitroanthranilic acid, 2-(Acetylamino)-6-nitrobenzoic Acid, N-Acetyl-6-nitroanthranilic Acid

A151551-1G A151551-250MG A151551-25G A151551-5G
ALA151551-1GEA 61.66 USD
ALA151551-1G ALA151551250MG ALA151551-25G ALA151551-5G
2-Acetamido-6-nitrobenzoic acid ≥98%
2-Acetamido-6-nitrobenzoic acid
Formula: C₉H₈N₂O₅
MW: 224.17 g/mol
Melting Pt: 214 °C
Storage Temperature: Ambient
MDL Number: MFCD08276267
CAS Number: 73721-78-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f51b9f3defb31f0f6b0950026e175002157221c8&vid=f51b9f3defb31f0f6b0950026e175002157221c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR