4-Pentylaniline 97 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amylaniline, 4-n-Pentylaniline, 4-Pentylaniline

A151044-25ML A151044-5ML
ALA151044-25MLEA 121.51 USD
ALA151044-25ML ALA151044-5ML
4-Pentylaniline 97 GC_ASSAY_METHOD
4-Pentylaniline

4-Pentylaniline was used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol.

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 265 °C
Flash Pt: >110°C
Storage Temperature: Ambient
MDL Number: MFCD00007926
CAS Number: 33228-44-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e42f91ecdf7aa49d3423339cfe8ba99a099348&vid=52e42f91ecdf7aa49d3423339cfe8ba99a099348&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR