2-Amino-3-chloropyridine 98 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Danger

A133692-100G A133692-1G A133692-5G
ALA133692-100GEA 246.91 USD
ALA133692-100G ALA133692-1G ALA133692-5G
2-Amino-3-chloropyridine 98 GC_ASSAY_METHOD
2-Amino-3-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56
Melting Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=113a662dcf2d253c099ae561e139769157cf0061&vid=113a662dcf2d253c099ae561e139769157cf0061&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR