3-Amino-4-fluorobenzoic acid ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Amino-4-fluorobenzoic Acid

A133076-1G A133076-5G
ALA133076-1GEA 19.86 USD
ALA133076-1G ALA133076-5G
3-Amino-4-fluorobenzoic acid ≥97% (by titration)
3-Amino-4-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00130055
CAS Number: 2365-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cca388774fef7c860a9e1e3c36f1cca8d8959dab&vid=cca388774fef7c860a9e1e3c36f1cca8d8959dab&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR