Isoquinoline-1-amine 97

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Isoquinolin-1-amine, 1-Aminoisoquinoline

A122683-5G A122683-1G A122683-25G
ALA122683-5GEA 378.01 USD
ALA122683-5G ALA122683-1G ALA122683-25G
Isoquinoline-1-amine 97
Isoquinoline-1-amine
Formula: C₉H₈N₂
MW: 144.18
Boiling Pt: 324
Melting Pt: 122-124°C
Density: 1.21
Storage Temperature: Ambient
MDL Number: MFCD00024137
CAS Number: 1532-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0705d79b24db9fa1a992f75f231e101505ba09e5&vid=0705d79b24db9fa1a992f75f231e101505ba09e5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR