Azido-PEG6-amine 98

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Azido-PEG-amine(n=6), O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol, Azido-PEG6-amine, N3-PEG6-CH2CH2NH2

A122194-100MG A122194-1G A122194-250MG A122194-5G
ALA122194100MGEA 93.96 USD
ALA122194100MG ALA122194-1G ALA122194250MG ALA122194-5G
Azido-PEG6-amine 98
Azido-PEG6-amine
Formula: C₁₄H₃₀N₄O₆
MW: 350.411 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03453240
CAS Number: 957486-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a668f6c2208eed64cf386810db4673f964e5804c&vid=a668f6c2208eed64cf386810db4673f964e5804c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR