Column Installation Pre-Swaging Tools for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
19251-80680 G3440-80217 G3440-80227 G3440-80218 G3440-80247
103577-048EA 100.05 USD
103577-048 103577-050 103577-052 AG19251-80680 AGG3440-80217 AGG3440-80227 AGG3440-80218 AGG3440-80247
Column Installation Pre-Swaging Tools for GC, Agilent Technologies
Puller and Inserter Tools
Handy stainless-steel tools for proper column installation lengths into the most popular GC inlets.
  • Available for Agilent 7890/6890 model split/split less, multimode, and purged packed inlets
  • Each tool has a fitting specific to its ferrule type to provide optimal sealing
  • one for graphite ferrules, one for metal ferrules
  • Easy to read markings for split/split less, multimode, and purged packed inlet lengths
  • A ferrule release hole on the preswaging tool for metal ferrules (G3440-80218) ensures against overtightening resulting in a stuck ferrule