Bond Elut LMS Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
A3961025
AGA3961025EA 548 USD
AGA3961025
Bond Elut LMS Plates, Agilent Technologies
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut LMS enables elution without the secondary reactions of additives.
  • Ultraclean styrene-divinylbenzene polymer
  • Optimized 75 µm particle size for reproducible flow
  • High capacity and surface area for efficient extraction
  • Large pore size (300 Å) for large molecule extraction
Bond Elut LMS (large molecule size) polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that analytes can be eluted with pure organic solvents or HPLC-compatible solvent mixtures of lowMore Product Information