CrossLab FTIR Instrument and Imaging System Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
899-1794 925-0128 883-9750 G8180-67610 PIKE-161-1903 PIKE-162-1320 SPECAC04090
AG899-1794PK 566 USD
AG899-1794 AG925-0128 AG883-9750 AGG8180-67610 AGPKE-161-1903 AGPKE-162-1320 AGSPECAC04090
CrossLab FTIR Instrument and Imaging System Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Optimize performance and maximize efficiency with Cary FTIR instrument and imaging system supplies.
  • Use the coarse gold mirror reference cap for standard diffuse samples or external reflectance
  • Use the smooth gold mirror reference cap for external reflectance (45° or grazing angle)
  • Use the coarse silver mirror reference cap for extremely diffuse samples
  • Use the smooth silver mirror reference cap for highly reflective samples typically requiring grazing angle measurements
An external reflectance measurement using FTIR requires a reference sample toMore Product Information