Bond Elut PBA Plates
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7541201C
101402-242EA 679.72 USD
101402-242 101403-188 101473-730 AG7541201C
Bond Elut PBA Plates
Microplates Solid Phase Extraction Plates (SPE)
Unique bond Elut PBA silica sorbent uses phenylboronic acid functionality to retain analytes via a reversible covalent bond, with high specificity and cleanliness.
  • Unique phenylboronic acid sorbent
  • High specificity for cis-diol compounds
  • Amenable for a broad range of biomolecule applications
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This very strong covalent retention mechanism enables high specificity and cleanliness.
The boronate groupMore Product Information