VersaPlate 96-Well-PPL
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
75480060 75409025
101473-748EA 484.23 USD
101473-748 103579-942 103579-946 103579-948 103579-952 103579-954 AG75480060 AG75409025
VersaPlate 96-Well-PPL
Microplates Solid Phase Extraction Plates (SPE)
VersaPlate allows you to process more samples in less time with less waste in a 96-well format.
  • Use only the number of wells you need for each run to save on sample preparation costs, while still enjoying all the benefits of automation
  • Increased well depth reduces the risk of overflow, and flangeless, beveled rims provide a tight fit. This helps avoid cross contamination
  • Scale up using the exact same product that was used with small scale development
Develop an SPE method faster, using liquid handling automation or multichannel pipettes, or useMore Product Information