Synthetic Chem Elut S Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5610-2003
AG5610-2003PK 247 USD
AG5610-2003
Synthetic Chem Elut S Plates, Agilent Technologies
Microplates Solid Supported Liquid/Liquid Extraction Plates (SLE)
Chem Elut S products offer superior SLE reproducibility with minimal method development.
  • Controlled particle and pore size enable consistent flow and reproducibility
  • Absence of shaking eliminates emulsion formation
  • Effective removal of matrix, including salts and phospholipids
  • Minimal extraction solvent required, leading to high analyte concentrations
  • Reduced analyst-to-analyst variability compared to conventional LLE
Chem Elut S products enable reproducible SLE results with high data quality and sample capacity. Consistent pore and particle sizesMore Product Information