Split/Splitless Inlet Gold Seals for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5182-9652 5188-5367 5190-2209 5190-6144 5190-6145 18740-20880 5190-6149
101401-026PK 375.25 USD
101401-026 101401-028 101471-152 101473-080 101480-474 101482-480 AG5182-9652 AG5188-5367 AG5190-2209 AG5190-6144 AG5190-6145 AG18740-20880 AG5190-6149
Split/Splitless Inlet Gold Seals for GC, Agilent Technologies
Seals Mechanical Seals
Using a patent pending manufacturing process, our new inlet gold seals eliminate tiny leaks from machining that can negatively impact your GC performance.
  • Improved production process gives smoother surface for better seal
  • Inert gold surface applied with a proven proprietary plating process
  • New kit includes washer – at no additional cost
  • Better packaging to ensure cleanliness
  • The new gold seal (5188-5367) is an enhancement of original gold seal (18740-20885)
The gold seal forms a leak-free seal with the bottom of the inlet body and column ferrule toMore Product Information