CrossLab Manual Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5190-1463 5190-1483 5190-1490 5190-1497 5190-1510 5190-1517 5190-1523 5190-1528 G4277-80120 5190-0464 5190-1464 5190-1465 5190-1471 5190-1479 5190-1487 5190-1488 5190-1489 5190-1491 5190-1493 5190-1498 5190-1500 5190-1503 5190-1507 5190-1529 5190-1530 5190-1531 5190-1532 5190-1457 5190-1460 5190-1466 5190-1472 5190-1475 5190-1476 5190-1481 5190-1495 5190-1502 5190-1504 5190-1506 5190-1509 5190-1511 5190-1513 5190-1514 5190-1516 5190-1518 5190-1519 5190-1521 5190-1524 5190-1525 5190-1527 5190-1533 5190-1534 5190-1535 5190-1536 5190-1537 5190-1539 5190-1540 5190-1541 5190-1542 5190-1543 5190-1544 5190-1545 5190-1547
AG5190-1463EA 109.3 USD
AG5190-1463 AG5190-1483 AG5190-1490 AG5190-1497 AG5190-1510 AG5190-1517 AG5190-1523 AG5190-1528 AGG4277-80120 AG5190-0464 AG5190-1464 AG5190-1465 AG5190-1471 AG5190-1479 AG5190-1487 AG5190-1488 AG5190-1489 AG5190-1491 AG5190-1493 AG5190-1498 AG5190-1500 AG5190-1503 AG5190-1507 AG5190-1529 AG5190-1530 AG5190-1531 AG5190-1532 AG5190-1457 AG5190-1460 AG5190-1466 AG5190-1472 AG5190-1475 AG5190-1476 AG5190-1481 AG5190-1495 AG5190-1502 AG5190-1504 AG5190-1506 AG5190-1509 AG5190-1511 AG5190-1513 AG5190-1514 AG5190-1516 AG5190-1518 AG5190-1519 AG5190-1521 AG5190-1524 AG5190-1525 AG5190-1527 AG5190-1533 AG5190-1534 AG5190-1535 AG5190-1536 AG5190-1537 AG5190-1539 AG5190-1540 AG5190-1541 AG5190-1542 AG5190-1543 AG5190-1544 AG5190-1545 AG5190-1547
CrossLab Manual Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes for manual injection can be used with any type or brand of manual injection valve.
  • Blunt-tip point style prevents damage to valve's internal parts
  • Can be used with any type or brand of manual injection valve
Agilent syringes for manual injection have a blunt-tip needle to prevent damaging the valve's internal parts. These syringes can be used with any type or brand of manual injection valve. For more help choosing the right syringe for your application, check out our syringe selection tool.