CrossLab Color Coded Syringes for Manual Injection Valves, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5190-1501 5190-1484 5190-1508 5190-1522 5190-1526 5190-1480 5190-1485 5190-1492 5190-1494 5190-1499 5190-1505 5190-1512 5190-1515 5190-1520
AG5190-1501EA 62.7 USD
AG5190-1501 AG5190-1484 AG5190-1508 AG5190-1522 AG5190-1526 AG5190-1480 AG5190-1485 AG5190-1492 AG5190-1494 AG5190-1499 AG5190-1505 AG5190-1512 AG5190-1515 AG5190-1520
CrossLab Color Coded Syringes for Manual Injection Valves, Agilent Technologies
Syringes Syringes for Chromatography
Many Agilent manual syringes for GC have color-coded barrels to facilitate volume selection.
  • The blunt-tip point style needle is designed to prevent damaging the valves internal parts
  • Can be used with any type/brand of manual injection valve
  • For fast, easy identification, each denomination (0.5 µl to 50 ml) features a distinct color boldly embossed on the glass barrel
  • Each syringe is designed with an easy-to-read volume scale and eco-friendly packaging that doubles as a syringe stand
For more help choosing the right syringe for your application, checkMore Product Information