Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5182-0714 5182-0715 5182-0716 5182-0864 5182-0866 5183-2068 5183-2070 5190-2278 9301-1377 5182-0551 5182-0553 5182-0554 5182-0556 5182-0865 5182-0867 5183-2067 5183-2069 5183-2071 5183-2072 5183-2083 5183-4448 5188-5369 5188-6535 5190-2280 5190-4034 8010-0198 8010-0199 8010-0542 8010-0543 5067-0246 5182-0555 5182-0557 5182-0558 5182-0732 5182-0733 5182-0736 5182-0737 5182-0868 5182-0869 5183-2080 5183-2081 5183-2082 5183-4429 5183-4450 5188-6536 5190-4031 5190-9062 5190-9063 5190-9100 5191-8150 8010-0010 8010-0414 8010-0415 8010-0417 8010-0426 392611535 5067-0247 5182-0735 5183-2079 5183-4428 5183-4432 5190-3187 5190-4030 5190-4038 5190-6186 8010-0012 8010-0425 5067-0205 5182-0734 5183-4433 5190-6115 5190-6118 5190-6184
101389-802PK 68.99 USD
101389-802 101389-804 101401-006 101404-256 101404-284 101407-704 101409-388 101409-390 101409-606 101409-608 101449-194 101449-198 101472-894 101473-434 101473-436 101473-470 101476-634 101477-230 101477-382 101477-384 101482-296 101482-298 101482-300 101482-302 101482-304 101482-306 101482-318 101482-350 101482-352 101482-354 101482-356 101482-358 101482-360 101482-422 101482-424 101482-426 101482-428 101482-430 101482-432 101482-582 101482-584 101482-586 101482-588 101482-590 101482-604 101482-606 101482-608 101482-610 101482-612 101482-626 101482-628 101482-630 101482-632 103582-804 103582-932 103583-920 103583-922 103584-486 103584-488 103584-494 103584-496 103584-498 103584-500 103584-504 AG5182-0714 AG5182-0715 AG5182-0716 AG5182-0864 AG5182-0866 AG5183-2068 AG5183-2070 AG5190-2278 AG9301-1377 AG5182-0551 AG5182-0553 AG5182-0554 AG5182-0556 AG5182-0865 AG5182-0867 AG5183-2067 AG5183-2069 AG5183-2071 AG5183-2072 AG5183-2083 AG5183-4448 AG5188-5369 AG5188-6535 AG5190-2280 AG5190-4034 AG8010-0198 AG8010-0199 AG8010-0542 AG8010-0543 AG5067-0246 AG5182-0555 AG5182-0557 AG5182-0558 AG5182-0732 AG5182-0733 AG5182-0736 AG5182-0737 AG5182-0868 AG5182-0869 AG5183-2080 AG5183-2081 AG5183-2082 AG5183-4429 AG5183-4450 AG5188-6536 AG5190-4031 AG5190-9062 AG5190-9063 AG5190-9100 AG5191-8150 AG8010-0010 AG8010-0414 AG8010-0415 AG8010-0417 AG8010-0426 AG392611535 AG5067-0247 AG5182-0735 AG5183-2079 AG5183-4428 AG5183-4432 AG5190-3187 AG5190-4030 AG5190-4038 AG5190-6186 AG8010-0012 AG8010-0425 AG5067-0205 AG5182-0734 AG5183-4433 AG5190-6115 AG5190-6118 AG5190-6184
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Vials
Vials with 2 ml wide opening are constructed from type 33 and type 51 COE borosilicate glass, with low metal content, to protect your sample from destabilizing or leaching.
  • 2 ml wide opening vials have a 40% larger opening than standard opening vials
  • Uniformly flat bottom ensures compatibility with inserts
  • Precision-formed neck for optimal robotic arm handling
  • Unique thread design for consistently secure seal
  • Optional ceramic write-on spot with fill marks
  • Rigorous quality assurance for dimensional consistency from lot to lot
The associated 9 mm screwMore Product Information