CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
27-2005 27-2010 27-2100 27-2105 27-2000 27-2007 27-2015 27-2103 27-2110 27-2115 27-2120 27-2200 27-2205 27-2210 27-2215 27-2310 27-2315
AG27-2005PK 305.3 USD
AG27-2005 AG27-2010 AG27-2100 AG27-2105 AG27-2000 AG27-2007 AG27-2015 AG27-2103 AG27-2110 AG27-2115 AG27-2120 AG27-2200 AG27-2205 AG27-2210 AG27-2215 AG27-2310 AG27-2315
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Dissolution Apparatus
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
  • Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Order Now
SPECIFICATIONS
Environmentally Preferable