CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
27-0100 27-0102 33-9041 27-0101 27-0103 27-0104 27-0106 27-0107 27-0108 27-0109 27-3000 27-3001 27-3002 27-8005 27-8020 27-8036 27-8600 27-8620 27-8621 27-8622 33-9007 33-9009 33-9012 33-9018 33-9022 33-9025 33-9029 33-9046 33-9048 33-9051 33-9052
AG27-0100EA 356 USD
AG27-0100 AG27-0102 AG33-9041 AG27-0101 AG27-0103 AG27-0104 AG27-0105 AG27-0106 AG27-0107 AG27-0108 AG27-0109 AG27-3000 AG27-3001 AG27-3002 AG27-8005 AG27-8020 AG27-8036 AG27-8600 AG27-8620 AG27-8621 AG27-8622 AG33-9007 AG33-9009 AG33-9012 AG33-9018 AG33-9022 AG33-9025 AG33-9029 AG33-9046 AG33-9048 AG33-9051 AG33-9052
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Dissolution Apparatus
Agilent can provide many different dosage form holders for the Apparatus 7.
  • Accessories for the reciprocating holder apparatus 7
  • Simulates the biorelevant conditions for the skin, gastrointestinal tract and tissue required for transdermal, oral drugs and implants with biorelevant temperature agitation rates and retention times
  • Accommodates a variety of dosage form holders including reciprocating disks, angled disks, spring holders, stent holders, and pointed rods
  • Supports typical volume configurations of 5 to 300 ml, depending on instrument