Other Operating Supplies Kits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
190025200
AG190025200KT 7201 USD
AG190025200
Other Operating Supplies Kits, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Typical supplies for up to 12 months use of VGA 77 and SIPS 10/20 used with flame AAS instruments.
  • Maintain accurate analysis
  • Save as much as 15% over the cost of purchasing supplies individually
  • Keep your instruments running for up to 12 months (depending on use) without worrying about reordering supplies
Agilent's spares kit provides the typical operating supplies required for moderate use of the VGA 77 and SIPS 10/20 accessories used with flame Atomic Absorption systems.