J&W Ultra Inert DB-BAC1 and DB-BAC2 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
123-9334UI 123-9434UI 125-9334UI
AG123-9334UIEA 852 USD
AG123-9334UI AG123-9434UI AG125-9334UI
J&W Ultra Inert DB-BAC1 and DB-BAC2 Columns, Agilent Technologies
Chromatography Columns
Agilent DB-BAC1 Ultra Inert and DB-BAC2 Ultra Inert columns are able to provide separation of all important blood alcohol compounds.
  • Superior solution for accurate blood alcohol quantification
  • Optimized resolution and baseline separation of critical blood alcohol peaks
  • Excellent peak shape and accurate integration of low-concentration compounds
  • Accurate identification of challenging polar analytes, even at trace levels
  • Each column is tested with a uniquely designed QC test mixture to verify column performance
  • Rapid separation in just over 2 minutes using
More Product Information