J&W DB-WAXetr Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
122-7332 122-7333 122-7362 123-7333 123-7334 125-7332 125-7334 122-7332-INT 122-7332E 122-7333E 122-7362E 122-7363 123-7312 123-7332 123-7354 123-7362 123-7363 123-7363E 123-7364 125-7312 125-7314 125-7332E 125-7333 125-7334E 125-7362 128-7323
AG122-7332EA 747 USD
AG122-7332 AG122-7333 AG122-7362 AG123-7333 AG123-7334 AG125-7332 AG125-7334 AG122-7332-INT AG122-7332E AG122-7333E AG122-7362E AG122-7363 AG123-7312 AG123-7332 AG123-7354 AG123-7362 AG123-7363 AG123-7363E AG123-7364 AG125-7312 AG125-7314 AG125-7332E AG125-7333 AG125-7334E AG125-7362 AG128-7323
J&W DB-WAXetr Columns, Agilent Technologies
Chromatography Columns
A high-polarity PEG column with an extended temperature range for alcohols, solvents, benzenes, styrenes, and organic acids.
  • Polyethylene glycol (PEG)
  • High polarity
  • Extended temperature range
  • Excellent column-to-column repeatability
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP phase G16
  • Tested against the strictest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SUPELCOWAX 10, SUPEROX II, CB-WAX, Stabilwax, BP-20, 007-CW, Carbowax, Rtx-WAX, ZB-WAX, ZB-WAX
More Product Information