J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
122-7032UI 122-7033UI 122-7062UI 122-7032UI-INT 122-7033UI-INT 123-7032UI 125-7032UI 122-7012UI 122-7063UI 122-7062UI-INT 123-7012UI 123-7062UI 123-7063UI 123-7032UI-INT 123-7033UI-INT 125-7012UI 125-7031UI 125-7037UI 128-7022UI
AG122-7032UIEA 802 USD
AG122-7032UI AG122-7033UI AG122-7062UI AG1227032UI AG1227033UI AG123-7032UI AG125-7032UI AG122-7012UI AG122-7063UI AG1227062UIINT AG123-7012UI AG123-7062UI AG123-7063UI AG1237032UIINT AG1237033UIINT AG125-7012UI AG125-7031UI AG125-7037UI AG128-7022UI
J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Chromatography Columns
Deliver the highest levels of PEG stationary phase inertness performance for a range of challenging active analytes.
  • Improved peakshape for challenging polar compounds with DB-WAX Ultra Inert
  • Reliable column-to-column inertness performance
  • Bonded polyethylene glycol (PEG) selectivity equivalent to DB-WAX
  • No method revalidation needed
  • High polarity
  • Equivalent to USP G16
  • Excellent performance for acidic compounds, including free fatty acids – no need for FFAP type column
  • Bonded and crosslinked phase is solvent rinseable and tolerates aqueous injections