CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12-5149V 27-5000 27-5100 27-5120 12-5000V 12-5035V 12-5152V 27-5005 27-5110 27-5130 33-9000 12-5000 12-5010 12-5010V 12-5033 12-5034 12-5035 12-5040 12-5050 12-5055 12-5145 12-5146 12-5147 12-5148 12-5149 12-5153 12-5157 12-5157V 12-5158 12-5158V 12-5159 12-5200 12-5500 12-5505 13-0010 27-6000 27-6050 33-9005
AG12-5149VEA 511.8 USD
AG12-5149V AG27-5000 AG27-5100 AG27-5120 AG12-5000V AG12-5035V AG12-5152V AG27-5005 AG27-5110 AG27-5130 AG33-9000 AG12-5000 AG12-5010 AG12-5010V AG12-5033 AG12-5034 AG12-5035 AG12-5040 AG12-5050 AG12-5055 AG12-5070 AG12-5120 AG12-5125 AG12-5145 AG12-5146 AG12-5147 AG12-5148 AG12-5149 AG12-5153 AG12-5157 AG12-5157V AG12-5158 AG12-5158V AG12-5159 AG12-5200 AG12-5500 AG12-5505 AG13-0010 AG27-6000 AG27-6050 AG33-9005
CrossLab Dissolution Vessels and Carriers, Agilent Technologies
Dissolution Apparatus
EaseAlign, TruCenter and TruAlign dissolution vessels are available in a variety of sizes.
  • Vessel volume sizes include 100 and 200 ml, and 1 and 2 L sizes
  • Low actinic-red, peak, and verified versions are available for select volumes
  • Verified dissolution vessels and other accessories are available to support conformance guidelines for mechanical qualification
  • Conversion kits are available to support small- and large-volume dissolution testing
Agilent provides a variety of dissolution vessels like TruAlign, TruCenter, or EaseAlign vessels. These areMore Product Information