Dissolution Enhancer Cells, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12-4000 12-4001 12-4005 12-4015 12-4020 12-4021 12-5170
AG12-4000EA 1088.5 USD
AG12-4000 AG12-4001 AG12-4005 AG12-4015 AG12-4020 AG12-4021 AG12-5170
Dissolution Enhancer Cells, Agilent Technologies
Dissolution Apparatus
The Enhancer (immersion) Cell is a PTFE cell with adjustable volume and a retaining cap.
  • Ideal for testing ointments, creams, and gels
  • Available in several surface areas, including 0.5, 2.0, and 4.0 cm²
  • Used with traditional dissolution apparatus and a 200 ml flat-bottom vessel or 1 L vessel
  • Meets SUPAC-Semisolid Guidance
  • Alignment tools specific to the surface area are sold individually
The Enhancer (immersion) Cell is a PTFE cell with adjustable volume and a retaining cap to eliminate air pockets trapped against the skin or artificial membrane thatMore Product Information