GS-Q Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
115-3432
AG115-3432EA 965 USD
AG115-3432
GS-Q Columns, Agilent Technologies
Chromatography Columns
GS-Q is a PLOT column useful for separating ethane, ethylene, and ethyne.
  • Porous divinylbenzene homopolymer
  • Polarity between that of Porapak-Q and Porapak-N
  • Separates ethane, ethylene, and ethyne (acetylene)
  • Not recommended for quantification of polar compounds
  • Minimal conditioning time (1 hour) required
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W GS-Q is useful for the separation of various light gas compounds, including ethane, ethylene,More Product Information