Blocking Reagents, Denhardt's Solution (100X), Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62416-AD J62416-AE
AAJ62416-ADEA 162 USD
AAJ62416-AD AAJ62416-AE
Blocking Reagents, Denhardt's Solution (100X), Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers

Denhardt's solution is used as a blocking reagent for northern or southern blots to prevent probe binding on open membrane space and non-target nucleic acids.


  • Contains 2% ficoll (type 400, pharmacia), 2% polyvinylpyrrolidone and 2% bovine serum albumin
  • Soluble in water


Denhardt's reagent is recommended for northern hybridization, hybridization of RNA, single copy southern hybridization or hybridization of DNA, immobilized on nylon membranes.

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical Form Liquid
MDL No. MFCD00717549
Storage Temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR