BSA Blocking Buffer, 5% in PBS, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61089-AK J61089-AP
AAJ61089-AKEA 216.89 USD
AAJ61089-AK AAJ61089-AP
BSA Blocking Buffer, 5% in PBS, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers

Liquid


Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Colorless

Order Now

SPECIFICATIONS

Environmentally Preferable
Appearance Colorless
Physical Form Liquid
Storage Temperature -30°C to -10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR