5-Fluoro-2-methylindole 95%

Supplier: Thermo Scientific Chemicals
Warning

H66121-03 H66121-06
AAH66121-03EA 75.6 USD
AAH66121-03 AAH66121-06
5-Fluoro-2-methylindole 95%
5-Fluoro-2-methylindole

Purity: 95% CAS No.: 399-72-4 Molecular Formula: C9H8FN Formula Weight: 149.17 Physical Form: Solid Melting Point: 98-101° Beilstein Reference: 116627 MDL No.: MFCD02093649

Formula: C9H8FN
MW: 149.17 g/mol
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD02093649
CAS Number: 399-72-4
EINECS: 609-764-5

Order Now

Specification Test Results

Assay Minimum 95

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR