(S)-2-Aminopropanamide hydrochloride 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

H63019-06 H63019-14 H63019-22
AAH63019-06EA 43.6 USD
AAH63019-06 AAH63019-14 AAH63019-22
(S)-2-Aminopropanamide hydrochloride 95%
(S)-2-Aminopropanamide hydrochloride

L-Alaninamide hydrochloride, 95%


Purity: 95% CAS No.: 33208-99-0 Molecular Formula: C3H8N2O.HCl Formula Weight: 124.57 Physical Form: Solid Melting Point: 212-217° dec Optical Rotation: +11°(c=1 in methanol) MDL No.: MFCD00066145

Formula: C₃H₉ClN₂O
MW: 124.57 g/mol
Melting Pt: 212…217 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066145
CAS Number: 33208-99-0

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR