Fmoc-L-homoleucine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Homoleu-OH, N-Fmoc-L-homoleucine, Fmoc-HoLeu-OH

H62270-03 H62270-MD
AAH62270-03EA 206 USD
AAH62270-03 AAH62270-MD
Fmoc-L-homoleucine 95%
Fmoc-L-homoleucine

N-Fmoc-L-homoleucine, 95%


Purity: 95% CAS No.: 180414-94-2 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Physical Form: Solid MDL No.: MFCD00270206

Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270206
CAS Number: 180414-94-2

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR