5-Bromo-2-fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

H61518-06
AAH61518-06EA 42.4 USD
AAH61518-06 AAH61518-14
5-Bromo-2-fluorophenol 98%
5-Bromo-2-fluorophenol

5-Bromo-2-fluorophenol, 98%

Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 53 °C/0.2 mm
Storage Temperature: Ambient
MDL Number: MFCD07782066
CAS Number: 112204-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR