1,2-Bis(trimethoxysilyl)ethane 96%

Supplier: Thermo Scientific Chemicals
Danger

H60850-AA H60850-AC
AAH60850-AAEA 52.14 USD
AAH60850-AA AAH60850-AC
1,2-Bis(trimethoxysilyl)ethane 96%
1,2-Bis(trimethoxysilyl)ethane

1,2-Bis(trimethoxysilyl)ethane, 96%


Purity: 96% CAS No.: 18406-41-2 Molecular Formula: C8H22O6Si2 Formula Weight: 270.43 Flash Point: 109°(228°F) Physical Form: Liquid Density: 1.073 Refractive Index: 1.409 Appearance: Colorless Boiling Point: 103-104° UN No.: 3382 MDL No.: MFCD00053672

Formula: C₈H₂₂O₆Si₂
MW: 270.428 g/mol
Boiling Pt: 103…104 °C
Density: 1.073
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053672
CAS Number: 18406-41-2
EINECS: 242-285-6
UN: 3382
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR