(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%

Supplier: Thermo Scientific Chemicals
Warning

H54922-03 H54922-MD
AAH54922-03EA 456 USD
AAH54922-03 AAH54922-MD
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid ≥98%, ee 99%
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid

Purity: 98% CAS No.: 96886-55-4 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Storage Temperature: 2°C to 8°C Melting Point: 285° Optical Rotation: -14° (c=1.3 in 1N HCl) MDL No.: MFCD00145248

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00145248
CAS Number: 96886-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR